Flowline LU10-1305 Ultrasonic Level Switch-PP, Short, 3/4" NPT
Representative image only.
Flowline Mfr # LU10-1305 – Item # EW-23002-14
Compatible with Smart Trak and Switch-Pak installation fittings
  • CSA approved intrinsically safe for use in hazardous environments
  • 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity
PRODUCT OPTIONS
$285.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Relay OutputSPST relay rated for 60 VA
  • Wetted MaterialPolypropylene
  • Max Process Temperature (° C)80
  • Min Process Temperature (° F)-40
  • Max Pressure (PSI)150
  • Power (VDC)12 to 36
  • DescriptionUltrasonic Level Switch-PP, Short, 3/4" NPT
Key Features
  • CSA approved intrinsically safe for use in hazardous environments
  • 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity
More About this Item
The LU10 series ultrasonic level switch is an excellent choice for a broad range of liquids, including those with light coating or scaling-type characteristics. Liquid examples include xylene, chromic acid and light cooking oil.