Flowline LU10-2305 Ultrasonic level switch; SPDT relay: 1 A at 60 VDC/VAC; 12-36 VDC

Representative image only.
Flowline Mfr # LU10-2305 – Item # EW-43299-06
Compatible with Smart Trak and Switch-Pak installation fittings
  • CSA approved intrinsically safe for use in hazardous environments
  • 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity

PRODUCT OPTIONS

$405.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy Of Depth Range±1 mm
  • Relay OutputSPDT relay: 1 A at 60 VAC/VDC
  • Max Process Temperature (° C)90
  • Min Process Temperature (° F)-40
  • Max Pressure (PSI)150
  • Power (VDC)12 to 36
  • DescriptionUltrasonic level switch; SPDT relay: 1 A at 60 VDC/VAC; 12-36 VDC

Key Features

  • CSA approved intrinsically safe for use in hazardous environments
  • 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity

More About this Item

The LU10 series ultrasonic level switch is an excellent choice for a broad range of liquids, including those with light coating or scaling-type characteristics. Liquid examples include xylene, chromic acid and light cooking oil.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat