Fluke 0.75X Wide-Angle Infrared Smart Lens
Representative image only.
Fluke Mfr # FLK-0.75X-WIDE-LENS – Item # EW-39749-50
You've already invested in a Fluke tool, now expand its capabilities with Fluke Genuine Accessories
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTi450 Wide Angle Lens
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These accessories increase the functionality of your test tool and increase your safety and proficiency. With Fluke Genuine Accessories, you can broaden the ability to take your Thermal Imager measurements in different environments to meet your application needs.
When working in a tight space, get a wide field of view that lets you see a larger target from a close distance.
Compatible with Fluke TiX580, TiX560, TiX520, TiX500, Ti480 PRO, Ti450 PRO, Ti450 SF6, Ti480, Ti450, Ti400 and Ti300 IR cameras.