Fluke 1760 BASIC Power Quality Recorder, No V or I Probes, No Trans
Representative image only.
Fluke Mfr # FLUKE-1760 BASIC – Item # EW-20035-75
Class A compliance for the most demanding power quality tests
  • Fully Clas A compliant
  • GPS time synchronization
  • Flexible and fully configurable thresholds and scale factors
  • Uninterrupted power supply (40 minutes)
  • 10 MHz, 6000 Vpk waveform capture
  • 2 GB data memory
  • Comprehensive software included
  • Plug and play rugged field design
  • 1760 Basic Includes
  • 8 input-channels (4 currents/4 voltages or 8 voltages)
  • Internal Flash-memory 2 GB
  • PQ Analyze software on CD-ROM
  • Ethernet cable for network connection (1)
  • Crosslink Ethernet cable for direct PC connection (1)
  • Mains cable (1)
  • Operational manuals
  • Carrying bag
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Max (Volts)6000
  • AC Max (Volts)6000
  • DescriptionPower Quality Recorder, No V or I Probes, No Trans
  • Warranty2 Years
Two year warranty
Key Features
  • Fully Clas A compliant
  • GPS time synchronization
  • Flexible and fully configurable thresholds and scale factors
  • Uninterrupted power supply (40 minutes)
  • 10 MHz, 6000 Vpk waveform capture
  • 2 GB data memory
  • Comprehensive software included
  • Plug and play rugged field design
  • 1760 Basic Includes
  • 8 input-channels (4 currents/4 voltages or 8 voltages)
  • Internal Flash-memory 2 GB
  • PQ Analyze software on CD-ROM
  • Ethernet cable for network connection (1)
  • Crosslink Ethernet cable for direct PC connection (1)
  • Mains cable (1)
  • Operational manuals
  • Carrying bag
  • 2 year warranty
More About this Item
The Fluke 1760 Three-Phase Power Quality Recorder is fully compliant to IEC 61000-4-30 Class-A, for advanced power quality analysis and consistent compliance testing. Designed for analysis of utility and industrial power distribution systems, in medium- and low-voltage networks, this power quality monitor provides the flexibility to customize thresholds, algorithms, and measurement selections. The 1760 power recorder captures the most comprehensive details on user-selected parameters.
Sold in the United States and Canada only