This item is now discontinued. We recommend the following as suggested replacements: EW-20050-90
FLUKE-33 true-rms AC clamp meter
Representative image only.
Fluke Mfr # FLUKE-33 – Item # EW-26033-00
Tapered jaws give you easy access to tight spaces4000-count LCD with 40-segment analog bar graph True-rms AC current measurements Sleep mode preserves battery life Display hold
SPECS & DESCRIPTION
Specifications & Description
  • AC Min (amps)40
  • AC Max (amps)700
  • AC Current Accuracy±2.0%, ±5% from 400 to 700 A
  • Frequency Min (Hz)0
  • Frequency Max (Hz)10
  • Descriptiontrue-rms AC clamp meter
More About this Item
Measure AC current and frequency of both nonlinear and traditional loads. Peak value of the current waveform indicates presence of harmonics. The Fluke ® 33 also stores minimum, maximum, and average readings.
Meter includes holster (Fluke® model no. C30) and one 9 V battery. Order test leads separately in the "Accessories" section (under the product picture).