Fluke 4917619 TI-Series Eyepiece
Representative image only.
Fluke Mfr # FLK-TI-EYEPIECE – Item # EW-39749-49
You've already invested in a Fluke tool, now expand its capabilities with Fluke Genuine Accessories
  • High resolution of 800x480
  • Easy 4K and Ultra HD support via HDMI
  • Enhanced communication with HDMI input and output
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTI-Series Eyepiece
Two year warranty
Key Features
  • High resolution of 800x480
  • Easy 4K and Ultra HD support via HDMI
  • Enhanced communication with HDMI input and output
  • 2 year warranty
More About this Item
These accessories increase the functionality of your test tool and increase your safety and proficiency. With Fluke Genuine Accessories, you can broaden the ability to take your Thermal Imager measurements in different environments to meet your application needs.
Compatible with Fluke Infrared Cameras, this electronic viewfinder is ideal for situations when there is glare on the LCD screen, when work conditions require awkward camera angles, or as a second monitor for view by multiple people of what the camera is capturing.