Fluke 700HTPK Hydraulic Test Pressure Kit

Representative image only.
Fluke Mfr # FLUKE-700HTPK – Item # EW-68450-95
Test pump, hose kit, and adapters bundled for convenience and value
  • Combine a hydraulic test kit with any Fluke 700G series gauge for a complete pressure testing solution. Each kit includes the hoses and adapters needed for connecting the test pump to the pressure device to be tested. The precision gauge you select (purchased separately) connects directly to the test pump. The kit is also compatible for use with the 700PXX series pressure modules and 71X series calibrators.
  • 1 year warranty

$2,116.99USD / Each
sign in for your price.

$207.00 - $292.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Min Pressure (PSI)0
  • Max Pressure (PSI)10000
  • Instrument Connection Npt(F) (in)1/4
  • Media CompatibilityDistilled water and hydraulic mineral oil
  • DescriptionHydraulic Test Pressure Kit
  • Warranty1 Year
1 Year Warranty

Key Features

  • Combine a hydraulic test kit with any Fluke 700G series gauge for a complete pressure testing solution. Each kit includes the hoses and adapters needed for connecting the test pump to the pressure device to be tested. The precision gauge you select (purchased separately) connects directly to the test pump. The kit is also compatible for use with the 700PXX series pressure modules and 71X series calibrators.
  • 1 year warranty

More About this Item

Designed to generate calibration pressures up to 10,000 psi (690 bar)
Works with any 700G precision test gauge
Uses distilled water or mineral-based hydraulic oil as a source for calibration pressure

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat