Fluke 707 Compact Current Loop Calibrator

Representative image only.
Fluke Mfr # FLUKE-707 – Item # EW-26071-90
  • Large knob allows for easy one-handed operation.
  • Use to troubleshoot and calibrate transmitters
  • Simultaneous mA and % readout; provides 24 V internal loop power
  • Includes protective holster (Fluke model C10) and alligator test clips (Fluke model AC70A)
  • 3 year warranty

$1,168.99USD / Each
sign in for your price.

$174.00 - $244.00USD / Each

for an additional
$174.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Width (in)2
  • Height (in)1
  • Length (in)6
  • Batteries1 x 9V battery
  • DescriptionCompact Current Loop Calibrator
  • Warranty3 Years
3 Year Warranty

Key Features

  • Large knob allows for easy one-handed operation.
  • Use to troubleshoot and calibrate transmitters
  • Simultaneous mA and % readout; provides 24 V internal loop power
  • Includes protective holster (Fluke model C10) and alligator test clips (Fluke model AC70A)
  • 3 year warranty

More About this Item

Rugged calibrators are EMI-tolerant, dust-resistant, and splash-resistant.

Available for sale in the United States and Canada only.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat