Fluke 717 30G Pressure Calibrator, 30 psig
Representative image only.
Fluke Mfr # FLUKE-717 30G – Item # EW-68450-84
  • 3 year warranty
PRODUCT OPTIONS
$2,086.99USD / Each
sign in for your price.
$200.00 - $270.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Operating Temperature Range−10 °C to 55 °C
  • Min Pressure-12 psi
  • Pressure Accuracy0.05% of range
  • DescriptionPressure Calibrator, 30 psig
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Pressure source and milliamp measurement capability to calibrate and maintain almost any pressure device with one tool
Error % calculation for quicker pass / fail decision making in the field
Best in class mA accuracy (0.015% for better e asuremetn confidence and workload coverage