Fluke 718 1G Pressure Calibrator, 1 psig
Representative image only.
Fluke Mfr # FLUKE-718 1G – Item # EW-68450-88
Best in class mA accuracy (0.015%) for better measurement confidence and workload coverage
  • Pressure source and milliamp measurement capability that calibrates and maintains almost any pressure device│Integrated pump with media protection│Error % calculation for quicker pass/fail decision making
PRODUCT OPTIONS
$2,759.99USD / Each
For better pricing please sign in.
$175.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Max Measurement (mA)24
  • Max Pressure1 psig
  • Min Pressure-1
  • Display Type5-digit LCD
  • BatteriesTwo 9V alkaline
  • Battery life4 to 20 hours
  • DescriptionPressure Calibrator, 1 psig
Key Features
  • Pressure source and milliamp measurement capability that calibrates and maintains almost any pressure device│Integrated pump with media protection│Error % calculation for quicker pass/fail decision making
More About this Item
The Fluke® 718 Pressure Calibrator provides a total pressure calibration solution for transmitters, gauges and switches. This compact pressure calibration solution is about a third of the size of comparable instruments and weighs just one kilogram (two pounds). The unique easy to clean pump helps protect the pump from damage and provides the ability to service the pump without disassembly, even in the field.