Fluke 721EX-1603 Pressure Calibrator 16 PSIG to 300 PSIG

Representative image only.
Fluke Mfr # FLK-721EX-1603 – Item # EW-16109-01
Provide pressure calibration and temperature measurement for custody transfer applications
  • Both pressure inputs perform pressure switch tests
  • Large backlit graphic LCD displays up to three inputs simultaneously
  • Store five instrument setups for quick recall of routine tests
  • Choose between standard and intrinsically safe models

PRODUCT OPTIONS

$3,421.99USD / Each
sign in for your price.

$216.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Max Measurement (VDC)24
  • Min Measurement (VDC)0
  • Max Measurement (mA)24
  • Min Measurement (mA)0
  • Max Pressure16 psig sensor 1, 300 psig sensor 2
  • Min Pressure-14 psig sensor 1, -12 psig sensor 2
  • Pressure Resolution0.01 psi (0.0001 bar) sensor 1, 0.1 psi (0.001 bar) sensor 2
  • Pressure Accuracy0.025% FS sensor 1, 0.025% FS sensor 2
  • Width (in)4
  • Height (in)2
  • Length (in)7
  • Width (cm)11
  • Height (cm)5.8
  • Length (cm)20
  • Batteries4 x AA batteries
  • DescriptionPressure Calibrator 16 PSIG to 300 PSIG

Includes

test leads, soft case, four AA batteries, and calibration document supplied by the manufacturer.

Key Features

  • Both pressure inputs perform pressure switch tests
  • Large backlit graphic LCD displays up to three inputs simultaneously
  • Store five instrument setups for quick recall of routine tests
  • Choose between standard and intrinsically safe models

More About this Item

Dual-isolated pressure sensors make the Fluke 721 precision calibrator the ideal tool for gas custody transfer applications, allowing you to take simultaneous static and differential pressure measurements with one calibrator.

Intrinsically safe 721Ex models (16109-00 through -14) are ATEX Zone I and IECEx rated for use in hazardous environments.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat