Fluke 721EX-3630 Pressure Calibrator 36 PSIG to 3000 PSIG

Representative image only.
Fluke Mfr # FLK-721EX-3630 – Item # EW-16109-13
Provide pressure calibration and temperature measurement for custody transfer applications
  • Both pressure inputs perform pressure switch tests
  • Large backlit graphic LCD displays up to three inputs simultaneously
  • Store five instrument setups for quick recall of routine tests
  • Choose between standard and intrinsically safe models
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Max Measurement (VDC)24
  • Min Measurement (VDC)0
  • Max Measurement (mA)24
  • Min Measurement (mA)0
  • Max Pressure36 psig sensor 1, 3000 psig sensor 2
  • Min Pressure-14 psig sensor 1, 0 psig sensor 2
  • Pressure Resolution0.01 psi (0.0001 bar) sensor 1, 0.1 psi (0.001 bar) sensor 2
  • Pressure Accuracy0.025% FS sensor 1, 0.025% FS sensor 2
  • Width (in)4 3/8
  • Height (in)2 3/8
  • Length (in)7 7/8
  • Width (cm)11
  • Height (cm)5.8
  • Length (cm)20
  • Batteries4 x AA batteries
  • DescriptionPressure Calibrator 36 PSIG to 3000 PSIG
  • Warranty3 Years
3 Year WarrantyIntrinsically Safe

Key Features

  • Both pressure inputs perform pressure switch tests
  • Large backlit graphic LCD displays up to three inputs simultaneously
  • Store five instrument setups for quick recall of routine tests
  • Choose between standard and intrinsically safe models
  • 3 year warranty

More About this Item

Dual-isolated pressure sensors make the Fluke 721 precision calibrator the ideal tool for gas custody transfer applications, allowing you to take simultaneous static and differential pressure measurements with one calibrator.

Intrinsically safe 721Ex models (16109-00 through -14) are ATEX Zone I and IECEx rated for use in hazardous environments.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat