Fluke A3003FC Wireless clamp meter, 2000 A DC
Representative image only.
Fluke Mfr # FLUKE-A3003FC – Item # EW-20049-57
See, save, and share data wirelessly — saving time and increasing productivity
  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWireless clamp meter, 2000 A DC
  • Warranty3 Years
3 Year Warranty
Key Features
  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 3 year warranty
More About this Item
The a3003 FC wireless 2000 A DC current clamp meter measures up to 2000 A DC and can be used as a standalone meter or as part of the Fluke connect system. Unit features a large jaw size (64 mm) for measuring large, high current conductors. The data logging function allows you to record and save up to 65,000 readings and in rush function provides additional flexibility.
The Fluke a3003 FC is a fully functional current clamp meter that wirelessly relays measurements to other Fluke Connect enabled master units including the Fluke 3000 FC DMM, Fluke Ti200/300/400, TiX 520/560 Infrared Cameras, and to your PC via optional PC3000 FC Adapter.
What’s included: Two AA batteries.
Available for sale in the United States and Canada only