Fluke C17XX Soft Carrying Case for Power Loggers
Representative image only.
Fluke Mfr # C17XX – Item # EW-20050-84
More visibility, reduced uncertainty, provides better power quality and energy consumption decisions
  • Automatically captures and logs voltage, current, power, harmonics and associated power quality values
  • Fluke Connect® compatible allows you to view data locally on the instrument or via Fluke Connect mobile app and desktop software
  • Power instrument directly from the measured circuit for added flexibility
  • 600 V CAT IV/1000 V CAT III highest rated safety in the industry for use at the service entrance and downstream
  • Measure all three phases and neutral with included 4 flexible current probes
  • Comprehensive logging allows more than twenty separate automatic logging sessions that can be easily stored on the instruments
  • Capture dips, swells, and inrush currents to help pinpoint potential root causes of power quality issues
  • Intuitive graphical setup allows you to capture the right data every time and reduces uncertainty about your connections with the intelligent verification function
  • In-the-field setup through the front panel or Fluke Connect App - no need to return to the workshop for download and setup or to take a computer to the electrical panel
  • Energy analyze plus application software allows you to easily download and analyze energy consumption and power quality state of health data
  • 2 year warranty
$188.09USD / Each
$208.99USD / Each
CONSIDER ALTERNATIVE…
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSoft Carrying Case for Power Loggers
  • Warranty2 Years
Two year warranty
Key Features
  • Automatically captures and logs voltage, current, power, harmonics and associated power quality values
  • Fluke Connect® compatible allows you to view data locally on the instrument or via Fluke Connect mobile app and desktop software
  • Power instrument directly from the measured circuit for added flexibility
  • 600 V CAT IV/1000 V CAT III highest rated safety in the industry for use at the service entrance and downstream
  • Measure all three phases and neutral with included 4 flexible current probes
  • Comprehensive logging allows more than twenty separate automatic logging sessions that can be easily stored on the instruments
  • Capture dips, swells, and inrush currents to help pinpoint potential root causes of power quality issues
  • Intuitive graphical setup allows you to capture the right data every time and reduces uncertainty about your connections with the intelligent verification function
  • In-the-field setup through the front panel or Fluke Connect App - no need to return to the workshop for download and setup or to take a computer to the electrical panel
  • Energy analyze plus application software allows you to easily download and analyze energy consumption and power quality state of health data
  • 2 year warranty
More About this Item
The Fluke 1736 three-phase power logger is designed with Fluke Connect® mobile app and desktop software compatibility providing you the data you need to make critical power quality and energy decisions in real-time. The ideal test tool for conducting energy studies and basic power quality logging, this unit automatically captures and logs over 500 power quality parameters giving you more visibility into data you need to optimize system reliability and savings.