Fluke Calibration 1523 Handheld Thermometer Model
Representative image only.
Fluke Calibration Mfr # 1523-156 – Item # EW-93202-50
The first standards thermometers to fit into the palm of your hand.
  • Visual trending and measurement
  • Simultaneously measures two channels
  • Reads PRTs, thermocouples, and thermistors
  • “Plug-and-play” calibration coefficients with SMART LEMO connector
  • Trends data graphically
  • Documents readings and statistics
  • 1 year warranty
PRODUCT OPTIONS
$2,658.00USD / Each
Time to recalibrate?
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeMulti-Parameter
  • Number Of Probe Channels1
  • Types Of Probes AcceptedThermocouple, Thermistor, RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)3182
  • Min Temperature (° C)-200
  • Max Temperature (° C)1750
  • Accuracy0.004% + 0.002 ohm percent of reading + floor
  • Resolution0.0001°
  • Height (in)7 7/8
  • Battery Life20 hours
  • Battery3 AA (included)
  • DescriptionHandheld Thermometer Model
  • Warranty1 year
1 Year WarrantyNIST Traceable Calibration
Key Features
  • Visual trending and measurement
  • Simultaneously measures two channels
  • Reads PRTs, thermocouples, and thermistors
  • “Plug-and-play” calibration coefficients with SMART LEMO connector
  • Trends data graphically
  • Documents readings and statistics
  • 1 year warranty
More About this Item
The 1523 and 1524 are not just basic hand held thermometers, but in fact are highly precise reference thermometers. These rugged thermometers are designed for engineers and technicians that want accuracy and versatility in an easy to use package. Use them with RTD's (PRTs), thermocouples, and thermistors probes to read, document, and visually trend temperature data.
Each unit uses a smart LEMO connector that automatically loads calibration coefficients unique to each probe for easy and accurate plug and play operation. (Only Fluke calibration probes with a "P" style Info-Con connector will fit this meter)
Two models are available distinguished by the number of channels they measure and data logging capability. Includes a free software program that makes it easy to download logged data and program new calibration coefficients.
Available for sale in the United States and Canada only.
Model 1523
Single channel that reads RTD's (PRTs), thermocouples, and thermistors. Featuring visual data trending and records the values and statistics associated with up to 25 measurements.