Fluke Calibration 1653875 PRT/RTD Bath Probe, 0.25" Dia x 12" L
Representative image only.
Fluke Calibration Mfr # 5626-12-I – Item # EW-90450-67
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)1228
  • Min Temperature (° C)-200
  • Max Temperature (° C)661
  • Temperature Accuracy±0.011°F (±0.006°C)
  • Connector TypeInfo-Con
  • Sheath Diameter (in)0.25
  • DescriptionPRT/RTD Bath Probe, 0.25\" Dia x 12\" L
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5626 Series - Secondary standard PRT/RTD probes: These probes fill the gap between affordable, but temperature-limited secondary PRTs and more expensive, highly accurate SPRTs. Using a regular PRT at temperatures above 500 °C exposes the platinum to contamination. If the PRT is used as a reference or calibration standard, contamination is a major problem. SPRTs, which are more expensive and delicate, can handle the higher temperatures, but with greater risk to the instrument due to shock, contamination, or mishandling. The 5626 is designed to reduce the contamination risk through the use of internal protection while not impairing performance The 5626 is nominally 100W.
Available for sale in the United States and Canada only.