Fluke Calibration 2019-DC8 Fluid Level Adapter Kit
Representative image only.
Fluke Calibration Mfr # 2019-DC8 – Item # EW-16103-32
  • 1 year warranty
PRODUCT OPTIONS
$2,766.79USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)115
  • DescriptionFluid Level Adapter Kit
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Hart's deep-well compact baths are small, fast and quiet, yet with enough depth to calibrate even the longest PRTs, SPRTs, and liquid-in-glass (LIG) thermometers.
  • Access opening 6.8 x 4.7 in; 18 in deep, or 19 in with optional fluid level adapter
  • RS232 ports for automated calibrations with optional MET / TEMP II software
  • Fast heat-up and cool-down times
  • Small, 4.2 gal (15.9L) fluid volume with expansion reservoir to catch fluid overflow
  • Optional LIG thermometer calibration kit and magnifying scope
Available for sale in the United States and Canada only.