Fluke Calibration 3622882 PRT/RTD Freezer Probe, 0.125" D x 2" L; Mini-Lug
Representative image only.
Fluke Calibration Mfr # 5606-50-L – Item # EW-90450-03
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)320
  • Min Temperature (° C)-200
  • Max Temperature (° C)160
  • Temperature Accuracy±0.072°F (±0.04°C)
  • Connector TypeMini Spade Lug
  • DescriptionPRT/RTD Freezer Probe, 0.125\" D x 2\" L; Mini-Lug
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5606 Series - Full immersion PRT/RTD probes: Designed to perform in extreme environments where both the transition junction and the lead wires are required to withstand temperatures covering the entire operating range of the probe. Applications include calibration or validation of sensors used in laboratory or bio freezers, walk-in refrigerators, autoclaves, ovens, stability test chambers, furnaces and incubators.
Available for sale in the United States and Canada only.