Fluke Calibration 5610-9-L Thermistor Probe 1/8" x 9" Sst
Representative image only.
Fluke Calibration Mfr # 5610-9-L – Item # EW-37852-31
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeThermistor
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature Accuracy±0.018°F (±0.01°C)
  • Connector TypeMini Spade Lug
  • DescriptionThermistor Probe 1/8\" x 9\" Sst
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5610 Series - Secondary thermistor probes: The small drift and low calibration uncertainty of only ±0.01°C makes this rugged stainless steel thermistor a wise choice when temperatures of interest are between 0 and 100°C. These probes are ideal for measuring temperature in air, liquid or soil.
Available for sale in the United States and Canada only.