Fluke Calibration 5615-9-L High Accuracy Temperature Probe
Representative image only.
Fluke Calibration Mfr # 5615-9-L – Item # EW-10508-69
  • 1 year warranty
PRODUCT OPTIONS
$1,647.00USD / Each
Time to recalibrate?
for an additional
$520.00USD / Each
for an additional
$593.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)788
  • Min Temperature (° C)-200
  • Max Temperature (° C)420
  • Temperature Accuracy±0.022°F (±0.012°C)
  • Connector TypeMini Spade Lug
  • Sheath Diameter (in)0.25
  • DescriptionHigh Accuracy Temperature Probe
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5615 Series - Secondary reference PRT/RTD probes
These probes are the workhorse of secondary calibration laboratories around the world. With a calibration uncertainty at 0°C of just ±0.012°C, the 5615 is a great reference standard for comparison calibrations in a bath or dry-well. The 5615 comes in your choice of 12, 9, or 6 inch sheath lengths.
The 12-inch and 9-inch probes range from –200 to 420°C, and the 6-inch covers the temperature range from –200 to 300°C. The 12-inch probes have a sheath diameter of 1⁄4", and the 9- and 6-inch probes have a sheath diameter of 3⁄16".
Available for sale in the United States and Canada only.