Fluke Calibration 5622-05-I Fast Response PRT Probe, 10" Info-Con
Representative image only.
Fluke Calibration Mfr # 5622-05-I – Item # EW-90449-26
$530.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeRTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)662
  • Min Temperature (° C)-200
  • Max Temperature (° C)350
  • Connector TypeInfo-Con
  • DescriptionFast Response RTD Probe
More About this Item
5622 Series - Fast response PRT/RTD probes
Designed for special temperature measurement applications requiring fast response or short immersion over a wide temperature range. These high-quality wire-wound sensors are small allowing heat to transfer to the sensors quickly, with a time constants as fast as 0.4 seconds. Immersion requirements for these probes is also a plus, ranging from just 10 mm to 64 mm (0.4 to 2.5 inch), depending on the model.
Available for sale in the United States and Canada only.