Fluke Calibration 5627A-9-D RTD Industrial Probe 9" x 3/16" OD
Representative image only.
Fluke Calibration Mfr # 5627A-9-D – Item # EW-90449-57
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypePRT/RTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)788
  • Min Temperature (° C)-200
  • Max Temperature (° C)420
  • Temperature Accuracy±0.089°F (±0.046°C)
  • Connector Type5-Pin DIN
  • DescriptionRTD Industrial Probe 9\" x 3/16\" OD
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5627A Series – Precision industrial PRT/RTD probes: One of the best features of this sensor is that it conforms to the standard 385 curve, letting you use your DIN/IEC RTD meters fully. Manufactured using a coil suspension element design for increased shock and vibration resistance. It has a mineral-insulated sheath with a minimum bend radius of 19 mm (3/4-inch) for flexibility and durability.
Available for sale in the United States and Canada only.