Fluke Calibration 6330 Compact Temperature Calibration Bath, 35C to 300C
Representative image only.
Fluke Calibration Mfr # 6330 – Item # EW-16101-45
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Temperature (° F)572
  • Min Temperature (° F)95
  • Power (VAC)115
  • DescriptionCompact Temperature Calibration Bath, 35C to 300C
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Model HR6330 bath delivers all the high temperature you need up to 300C. With stability and uniformity at 300C better than ± 0.015C and ± 0.020C respectively, calibrations can easily be performed at this high temperature with total uncertainty better than ± 0.05C. At lower temperatures, stability and uniformity are even better. The HR6330 is only 12 inches wide and less than 19 inches tall, so it fits easily onto a benchtop without consuming precious space.
Model HR7320 and HR7340 baths cover your needs for low temperature calibrations. The HR7320 covers a range from -20C to 150C and the HR7340 reaches even colder temperatures to -40C. Below 0C, these baths maintain an impressive stability of ± 0.005C with uniformities also better than ± 0.005C. No utility bath performs as well below 0C or at critical room and body temperatures - or even at important higher temperatures such as 100C and 122C.
For ultra cold temperatures, the HR7380 reaches -80C quickly and maintains a two sigma stability of ± 0.006C when it gets there. The HR7380 is a true metrology bath, not a chiller or circulator. With uniformity to ± 0.008C, comparison calibration of temperature devices can be performed with high precision.
Each bath includes an RS232 serial interface and HR9930 interface-it software for controlling your bath from a PC. With a Hart Scientific thermometer readout, such as a Black Stack, and the Met Temp II software, automated calibrations can run unattended.