Fluke Calibration 9143A Field Metrology Well; 37 to 662F/Insert A
Representative image only.
Fluke Calibration Mfr # 9143-A-156 – Item # EW-16101-75
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Temperature (° F)662
  • Min Temperature (° F)91
  • DescriptionField Metrology Well; 37 to 662F/Insert A
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
  • Easy to use
  • Large LCD display
  • Function keys and easy to navigate menus
  • Pre-programmed calibration routines
  • One-button push to set the temperature
  • Reference readout accepts "plug-n-play" smart connector
Field Metrology Wells offer accuracy, portability, and speed
for field calibration applications. These units are packed with
functionality and are remarkably easy to use. Field
Metrology Wells are lightweight, small, and quick to reach
temperature set points; yet they are stabile, uniform, and
precise. This industrial product line is perfect for transmitter-
loop calibration, comparison calibration, or a simple check
of a thermocouple sensor. With the "process" option, you have
no need to have to carry additional tools into the field. It includes
a built-in readout for resistance, voltage, and mA measurement;
24V loop power; and on-board documentation.
Available for sale in the United States and Canada only.