Fluke Networks CLIP-SET RJ45 to 8-Clip Test Lead
Representative image only.
Fluke Networks Mfr # CLIP-SET – Item # EW-30002-11
Quick network connectivity and copper/fiber cabling answers for frontline technicians
  • Link at 10/100/Gig – identify device capabilities and confirm actual speed/duplex status on copper or fiber links
  • Ping key devices – measure response of DHCP, router, DNS, and other devices or URLs
  • Verify 802.1X settings – authenticate with EAP type, user name, certificates, and password
  • Detect PoE service – verify voltage level, and pairs to IEEE 802.3af specifications
  • Identify switch port – supports Cisco, Extreme, and IEEE Link Layer Discovery Protocols
  • Document results – upload link status reports with USB port for faster problem resolution
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRJ45 to 8-Clip Test Lead
  • Warranty3 Months
3 Month Warranty
Key Features
  • Link at 10/100/Gig – identify device capabilities and confirm actual speed/duplex status on copper or fiber links
  • Ping key devices – measure response of DHCP, router, DNS, and other devices or URLs
  • Verify 802.1X settings – authenticate with EAP type, user name, certificates, and password
  • Detect PoE service – verify voltage level, and pairs to IEEE 802.3af specifications
  • Identify switch port – supports Cisco, Extreme, and IEEE Link Layer Discovery Protocols
  • Document results – upload link status reports with USB port for faster problem resolution
  • 3 month warranty
More About this Item
Choose LinkRunner Duo for 10/100/Gig copper and just add an SFP transceiver for 100/Gig fiber. Verify link speed and duplex status, ping up to 10 key devices, detect Power over Ethernet (PoE), resolve 802.1X security conflicts, verify cable wire map, and identify the nearest switch port with Cisco, Extreme and IEEE link layer discovery protocols (CDP/EDP/LLDP).
Built-in IntelliTone digital toning precisely locates copper cables at patch panels and switches. Generate professional link status test reports to expedite trouble-ticket resolution.