Fluke ES-162P3 3-Pole Fall of Potential Stake Kit
Representative image only.
Fluke Mfr # ES-162P3 – Item # EW-23031-86
Optimize performance of your ground resistance Meter Accessories
  • Needed to perform 3-Pole Fall of Potential testing (Stake testing)
SPECS & DESCRIPTION
Specifications & Description
  • Description3-Pole Fall of Potential Stake Kit
Key Features
  • Needed to perform 3-Pole Fall of Potential testing (Stake testing)
More About this Item
Suitable for use with both the Fluke 1623 and Fluke 1625 meters. Kit Includes: 3 ground/earth stakes, a 50m cable reel of wire and a 25m cable reel of wire.