Fluke FLK-A3000 FC Kit Wireless Kit with Current Module
Representative image only.
Fluke Mfr # FLK-A3000 FC KIT – Item # EW-20047-88
Economical kits save you time and money!
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Conduct direct or remote AC current measurements
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • Easily set up clamp inside the panel and view measurement s remotely at your convenience
PRODUCT OPTIONS
$386.99USD / Each
$429.99USD / Each
$65.00 - $152.80USD / Each
for an additional
$65.00USD / Each
for an additional
$95.00USD / Each
for an additional
$152.80USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWireless Kit with Current Module
Key Features
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Conduct direct or remote AC current measurements
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • Easily set up clamp inside the panel and view measurement s remotely at your convenience
  • AC module measures up to 400 A AC for added flexibility
More About this Item
These Fluke connect kits allow you to connect wirelessly to over 20 different Fluke products allowing you to share data without leaving the field. Kits combine wireless modules that allow you to monitor and maintain equipment to keep your business running smoothly.
This Fluke A3000 FC wireless digital multimeter kit with AC current clamp module provides you maximum flexibility for taking measurements for all of your application requirements. Jaw opening of 34 mm accommodates large cables and the wireless system allows you to easily set up measurements once and read them as often and when you want.
Available for sale in USA & Canada only
Whats included:
FC 3000 wireless digital multimeter, A3000 FC AC current module, TL175 test lead set, AC175 alligator clip set, C3003 Soft carrying case, three compartment.
Available for sale in the United States and Canada only