Fluke FLK-A3001 FC Kit Wireless Kit with Flexible Current Module
Representative image only.
Fluke Mfr # FLK-A3001 FC KIT – Item # EW-20047-89
Economical kits save you time and money!
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Basic wireless kit is ideal for remote AC current measurement applications
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • AC current clamp module with flexible probe measures up to 2500 A AC
  • Safely and accurately make AC current measurements from a distance
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWireless Kit with Flexible Current Module
  • Warranty3 Years
3 Year WarrantyConformite EuropeenneCanadian Standards AssociationIP54 Rated
Key Features
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Basic wireless kit is ideal for remote AC current measurement applications
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • AC current clamp module with flexible probe measures up to 2500 A AC
  • Safely and accurately make AC current measurements from a distance
  • 3 year warranty
More About this Item
These Fluke connect kits allow you to connect wirelessly to over 20 different Fluke products allowing you to share data without leaving the field. Kits combine wireless modules that allow you to monitor and maintain equipment to keep your business running smoothly.
This Fluke A30001 FC wireless digital multimeter kit with AC current clamp module and flexible probe provides you maximum flexibility for taking measurements for all of your application requirements. Wireless system allows you to easily set up measurements once and read them as often and when you want.
Available for sale in USA & Canada only
Whats included:
FC 3000 wireless digital multimeter, A3001 FC AC current module, iFlex 10” flexible AC current probe,
TL175 test lead set, AC175 alligator clip set, C3003 Soft carrying case, three compartment, magnetic hanging strap for hands-free operation.
Available for sale in the United States and Canada only