Fluke FLK-LENS/WIDE2 Thermal Imager Wide-Angle Lens, 2x
Representative image only.
Fluke Mfr # FLK-LENS/WIDE2 – Item # EW-39750-39
You've already invested in a Fluke tool, now expand its capabilities with Fluke Genuine Accessories
  • Field of view 46in x 34in
  • Minimum focus distance 3in
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionThermal Imager Wide-Angle Lens
  • Warranty1 year
1 Year Warranty
Key Features
  • Field of view 46in x 34in
  • Minimum focus distance 3in
  • 1 year warranty
More About this Item
These accessories increase the functionality of your test tool and increase your safety and proficiency. With Fluke Genuine Accessories, you can broaden the ability to take your Thermal Imager measurements in different environments to meet your application needs.