Fluke FLK-PC3000 FC Wireless PC Adapter Module
Representative image only.
Fluke Mfr # FLK-PC3000 FC – Item # EW-20047-82
See, save, and share data wirelessly — saving time and increasing productivity
  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 1 year warranty
PRODUCT OPTIONS
$121.49USD / Each
$134.99USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWireless PC Adapter Module
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 1 year warranty
More About this Item
The Fluke PC3000 FC Adapter and software help you understand the information you measure. When you need to get information back to your PC the PC3000 FC adapter and software wirelessly collects up to 65,000 sets of time stamped min/max/avg readings from Fluke Connect FC remote modules. The software can display live readings from up to six modules simultaneously and allows custom logging intervals and custom names for each module. Includes Windows® SW3000 software.
Software requirements:
The Fluke PC3000 must be connected to the USB port in order to enable communication between FC modules and software. SW3000 will support up to two PC3000FC adapters.
Operating system compatibility:
Windows XP or later.
Available for sale in the United States and Canada only