Fluke FLK-V3000 FC Kit Wireless Kit with AC Voltage Module
Representative image only.
Fluke Mfr # FLK-V3000 FC KIT – Item # EW-20047-91
Economical kits save you time and money!
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Monitor AC voltage and troubleshoot equipment remotely
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • Set up AC voltage measurements from a distance and view your convenience
  • Cat IV 600 V, CAT III 1000 V rated to ensure operator safety
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWireless Kit with AC Voltage Module
  • Warranty3 Years
3 Year WarrantyConformite EuropeenneCanadian Standards AssociationIP54 Rated
Key Features
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Monitor AC voltage and troubleshoot equipment remotely
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • Set up AC voltage measurements from a distance and view your convenience
  • Cat IV 600 V, CAT III 1000 V rated to ensure operator safety
  • 3 year warranty
More About this Item
These Fluke connect kits allow you to connect wirelessly to over 20 different Fluke products allowing you to share data without leaving the field. Kits combine wireless modules that allow you to monitor and maintain equipment to keep your business running smoothly.
This V3000 FC wireless digital multimeter kit with AC voltage module provides you maximum flexibility for taking measurements for all of your application requirements. Easily view measurements directly on the AC voltage module or remotely from the wireless digital multimeter for added flexibility.
Available for sale in USA & Canada only
Whats included
FC 3000 wireless digital multimeter, V3000 FC AC voltage module, TL175 and TL224 test lead sets,
AC175 and AC285 alligator clip sets, C3003 Soft carrying case, three compartment, magnetic hanging strap for hands-free operation.
Available for sale in the United States and Canada only