Fluke FLK-V3001 FC Kit Wireless Kit with DC Voltage Module
Representative image only.
Fluke Mfr # FLK-V3001 FC KIT – Item # EW-20047-92
Economical kits save you time and money!
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Conduct direct or remote DC voltage measurements
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • Easily set up clamp inside the panel and view measurements remotely at your convenience
PRODUCT OPTIONS
$395.99USD / Each
$439.99USD / Each
$65.00 - $94.50USD / Each
for an additional
$65.00USD / Each
for an additional
$94.50USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWireless Kit with DC Voltage Module
Key Features
  • Kits feature wireless capability for quickly and easily sharing data
  • Use with Fluke Connect mobile app for Android™ and iOS devices
  • Conduct direct or remote DC voltage measurements
  • See, save, and share data wirelessly with Fluke Connect mobile app for Android™ and iOS devices
  • Easily set up clamp inside the panel and view measurements remotely at your convenience
  • Logging function allows you to record and save up to 65,000 readings for expanded analysis capabilities
More About this Item
These Fluke connect kits allow you to connect wirelessly to over 20 different Fluke products allowing you to share data without leaving the field. Kits combine wireless modules that allow you to monitor and maintain equipment to keep your business running smoothly.
This Fluke V3001 FC wireless digital multimeter kit with DC voltage clamp module provides you maximum flexibility for taking measurements for all of your application requirements. Jaw opening of 34 mm accommodates large cables and the wireless system allows you to easily set up measurements once and read them as often and when you want.
Available for sale in USA & Canada only
Whats included
FC 3000 wireless digital multimeter, V3001 FC DC voltage module, TL175 and TL222 test lead sets,
AC175 and AC220 alligator clip sets, C3003 Soft carrying case, three compartment, one magnetic hanging strap for hands-free operation.
Available for sale in the United States and Canada only