Fluke FP PLUS Fp Plus Food Pro Plus Food Safety Infrared (IR) Thermometer
Representative image only.
Fluke Mfr # FLUKE-FP PLUS – Item # EW-39753-81
Complete temperature measurement solution for continual temperature monitoring
  • Surface temperature readings in less than half a second for quick temperature readings
  • HACCP check trigger safe/unsafe lights for instant
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Distance To Target Size Ratio (D:S)2.5:1
  • EmissivityFixed
  • Sighting TypeSingle-point laser
  • Min Temperature (° F)-30
  • Max Temperature (° F)525
  • Min Temperature (° C)-35
  • Max Temperature (° C)275
  • Accuracy±1.0°C; ±0.1% of reading ± 1.0°C below 0°C; ±1.5% of reading above 65°C
  • Resolution0.1°
  • Response time<500 milliseconds
  • Probe Min Temperature (° F)-40
  • Probe Max Temperature (° F)392
  • Probe Min Temperature (° C)-40
  • Probe Max Temperature (° C)200
  • Battery9v Battery (1)
  • Temperature InputYes
  • DescriptionFp Plus Food Pro Plus Food Safety Infrared (IR) Thermometer
  • Warranty1 year
1 Year Warranty
Key Features
  • Surface temperature readings in less than half a second for quick temperature readings
  • HACCP check trigger safe/unsafe lights for instant
  • 1 year warranty
More About this Item
The FoodPro series provides the total temperature solution for monitoring food temperatures.
The FoodPro model takes quick surface temperature scans with an infrared thermometer for instant temperature readouts without fear of cross-contamination.
A combination infrared and probe thermometer, the FoodPro Plus takes quick surface scans and internal temperatures all with one unit.
Used together, the FoodPro and FoodPro Plus units provide a complete temperature measurement solution for continual temperature monitoring.
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
We now offer an FDA-cleared infrared thermometer.
Our other infrared thermometers are not approved for medical usage and are not FDA-approved.
For help in selecting an infrared thermometer please contact the Technical Support team at Cole-Parmer.