Fluke Magnetic Hanger Kit for 700G Pressure Gauges
Representative image only.
Fluke Mfr # FLUKE-GAUGEPAK – Item # EW-20051-13
Designed to integrate seamlessly with the existing 700G Series protective holster, the hanger kit is both easy-to-install, and easy-to-hang, freeing up your hands for pumping pressure, taking readings, and documenting test and calibration results
  • Strong magnetic hanger strap for Fluke 700G Pressure Gauges
  • Hang with magnet or hook strap for non-magnetic surfaces
  • Fast, easy installation using the 700G Gauge's protective holster
  • Allows you to read pressure measurements hands free
  • 1 year warranty
$86.39USD / Each
$95.99USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMagnetic Hanger Kit for 700G Pressure Gauges
1 Year Warranty
Includes
Gauge hanger, magnetic hanger kit, and instruction card.
Key Features
  • Strong magnetic hanger strap for Fluke 700G Pressure Gauges
  • Hang with magnet or hook strap for non-magnetic surfaces
  • Fast, easy installation using the 700G Gauge's protective holster
  • Allows you to read pressure measurements hands free
  • 1 year warranty
More About this Item
The Fluke GAUGEPAK Magnetic Hanger Kit makes it easier than ever to take pressure measurements with a Fluke 700G Series Pressure Gauge. If you are looking to attach your pressure gauge to a metallic surface such as a nail, pipe, pump, or panel the strong rare-earth magnet will ensure that your gauge stays in place and is at the optimal viewing angle. When metallic surfaces aren’t available, the included hook-and-loop strap allows you to conveniently hang your gauge on a pole or pipe.