Fluke Networks CIQ-100 CableIQ Qualification Tester
Representative image only.
Fluke Networks Mfr # CIQ-100 – Item # EW-20021-84
Troubleshoot connectivity problems
  • Sees if your existing cabling can support voice, VoIP, 10/100/Gigabit Ethernet
  • Troubleshoots why existing cabling plant can’t support the desired network bandwidth
  • Discovers what’s at the end of any cable and displays device configuration (speed/duplex/pairing)
  • Frees-up switch ports and identifies unused links
  • Graphically maps wiring configuration and shows distance to faults with Intelligent Wiremap
  • Tests all copper cabling media: twisted-pair, coax, and audio cabling
  • Locates and traces cable with IntelliTone™ digital signaling technology
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCableIQ Qualification Tester
  • Warranty1 Year
1 Year Warranty
Key Features
  • Sees if your existing cabling can support voice, VoIP, 10/100/Gigabit Ethernet
  • Troubleshoots why existing cabling plant can’t support the desired network bandwidth
  • Discovers what’s at the end of any cable and displays device configuration (speed/duplex/pairing)
  • Frees-up switch ports and identifies unused links
  • Graphically maps wiring configuration and shows distance to faults with Intelligent Wiremap
  • Tests all copper cabling media: twisted-pair, coax, and audio cabling
  • Locates and traces cable with IntelliTone™ digital signaling technology
  • 1 year warranty
More About this Item
CableIQ is the first qualification tool designed for network technicians who need to troubleshoot connectivity problems and qualify cabling bandwidth. It will help you to determine if your cabling is qualifiedto support 10/100, VoIP or Gigabit Ethernet. Detects speed/duplex settings of attached switches and PCs. Intelligent wiremap graphically shows distance to faults. Intuitive, lightweight, and rugged.
CableIQ Qualification Tester includes CableIQ with remote adapter, CableIQ Reporter software CD, (2) RJ45-RJ45 patch cords, USB cable, coax 'F' push-on adapter, owners CD and quick start guide, (4) AA batteries, and soft carry case