Fluke TL222 Silicone Insulated Right-Angle Test Leads
Representative image only.
Fluke Mfr # TL222 – Item # EW-26002-32
High performance at an economical price
  • DMM test leads (red, black) with safety shrouded, standard diameter banana plugs
  • Right angle connector on both ends
  • Reinforced strain relief
  • 1.5 meter silicone-insulated wire resists heat and cold
  • CAT IV 600 V, CAT III 1000 V, 10 A rating. UL listed
  • 1 year warranty
PRODUCT OPTIONS
$39.59USD / Set of 1
$43.99USD / Set of 1
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSilicone Insulated Right-Angle Test Leads
  • Warranty1 Year
1 Year Warranty
Key Features
  • DMM test leads (red, black) with safety shrouded, standard diameter banana plugs
  • Right angle connector on both ends
  • Reinforced strain relief
  • 1.5 meter silicone-insulated wire resists heat and cold
  • CAT IV 600 V, CAT III 1000 V, 10 A rating. UL listed
  • 1 year warranty
More About this Item
Various test leads are available to suit your application.
Available for sale in the United States and Canada only.