FMI Series RH Valveless Piston Pump Head, Ceramic, 1/4", 1/4-28 (F) ports
Representative image only.
FMI Mfr # RH1CKCLF – Item # EW-07117-69
$619.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (mL/min)75
  • Min Flow Rate (mL/min)0.03
  • Max Pressure (PSI)100
  • Max Motor RPM (rpm)750
  • Min Motor RPM (rpm)6
  • Wetted MaterialsCeramic (alumina), PVDF
  • Inlet Connection1/4-28 female
  • Diameter (in)1/4
  • DescriptionLow Flow Pump Head for Q Series of Pumps, 1/4" Ceramic Piston dia
More About this Item
Valveless piston pumps are low maintenance and suffer fewer piston siezures or clogs. With a single moving part, there is less drift over time, offering millions of cycles of reliable, repeatable pumping.