FMI Series RH Valveless Piston Pump Head, Ceramic/PVDF/Ceramic, 1/4-28 NPT(F); 0-0.05 mL/Stroke
Representative image only.
FMI Mfr # FMI013 / RH0CKC-LF – Item # EW-07117-59
Small displacement provides excellent precision
  • Fully adjustable from zero to maximum
PRODUCT OPTIONS
$704.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Flow Rate (mL/min)37.5
  • Min Flow Rate (mL/min)0.018
  • Max Pressure (PSI)100
  • Max Motor RPM (rpm)750
  • Min Motor RPM (rpm)6
  • Wetted MaterialsCeramic (alumina), PVDF
  • Inlet Connection1/4-28 female
  • Diameter (in)3/16
  • DescriptionValveless Piston Pump Head, Ceramic/PVDF/Ceramic; 0 to 0.05 mL per Stroke, with 1/4-28 NPT(F) Ports
Key Features
  • Fully adjustable from zero to maximum
More About this Item
Valveless piston pumps are low maintenance and suffer fewer piston siezures or clogs. With a single moving part, there is less drift over time, offering millions of cycles of reliable, repeatable pumping.