Fowler 52-265-024 24in/600MM Rigid Pre-Cert. Rule
Representative image only.
Fowler Mfr # 522650240 – Item # EW-97604-59
Certified Rulers and Sets
  • 1 year warranty
PRODUCT OPTIONS
$188.00USD / Each
$46.80 - $120.00USD / Each
for an additional
$46.80USD / Each
for an additional
$120.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Description24in/600MM Rigid Pre-Cert. Rule
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
24"/600mm rigid ruler
Graduations in 32nds, 64ths, 1mm, and 0.5mm
Includes NIST-traceable calibration report