Fowler 52-438-777 gauge Ball Set Metric 52 Pc. Set
Representative image only.
Fowler Mfr # 52-438-777 – Item # EW-97604-17
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Mass CertificationAFBMA certificate
  • Descriptiongauge Ball Set Metric 52 Pc. Set
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Premium Brand 1-2-3 Blocks
  • Case hardened (HRC 55—60)
  • 23 holes for easy clamping
  • Useful in layout & grinding work
  • Squareness on all sides to within .0001"
  • Factory NIST certification available upon request. Add $80.00
Premium Brand 1/32" Parallel Set
  • Made from high-grade alloy steel, hardened and ground from 1/32" stock with 2 holes in each parallel.
  • 10 pairs, 6" length, 1/32" width
  • 1/2", 5/8", 3/4", 7/8", 1", 1-1/8", 1-1/4", 1-3/8", 1-1/2" & 1-5/8"
  • Height specifications relate to consistency within each pair. Absolute heights are nominal
  • Parallelism: ±.0001"
  • Paired in height: ±.0001"
  • Hardness: 50HRC
  • Factory NIST certification available upon request. Add $80.00
      gauge Ball Sets
      • Ball gauges are ground and lapped chrome steel
      • Hardened to 58 to 63RC.
      • Grade 24 accuracy-AFBMA Certificate
      • T52-438-766 Set includes 26 pairs
      • Pair sizes in inches: 1/8, 9/64, 5/32, 3/16, 7/32, 1/4, 9/32, 5/16, 11/32, 3/8, 13/32, 7/16, 15/32, 1/2, 17/32, 9/16, 19/32, 5/8, 21/32, 11/16, 23/32, 3/4, 13/16, 7/8, 15/16 and 1"
      • T52-438-777 Metric Set includes 26 pairs
      • Pair sizes in mm: 1mm to 25mm
      • Factory NIST certification available upon request. Add $430.00 Add $415.00 for metric
      12 Piece Angle Block Set