Fowler 52-664-609 Pocket Optical Comparator Set Comparator
Representative image only.
Fowler Mfr # 526646090 BOM – Item # EW-97604-13
  • 1 year warranty
$242.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Magnification10x
  • DescriptionPocket Optical Comparator Set Comparator
  • Warranty1 year
1 Year Warranty
Includes
optical comparator, illuminator, nine reticles, and durable wooden case.
Key Features
  • 1 year warranty
More About this Item
Fowler's 10x pocket optical comparator set produces magnified images which are sharp, undistorted, and aberration-free. Convenient for checking angles, thread measurements, hole sizes, as well as photo engraving, etching, and many more applications. The illuminator attachment assists where additional light is required.