Fowler 54-562-777 1/2" Electronic Test Indicator (Depth Gauges/Indicators)
Representative image only.
Fowler Mfr # 54-562-777 – Item # EW-10508-52
Fowler's QuadraTest offers unrivaled measuring versatility in an Electronic Test Indicator.
  • 4 measuring modes: Min, Max, TIR, and Direct
  • True inch / metric conversion
  • Water, oil and coolant resistant to IP65
  • 1-1/2" diameter dial face rotates a full 360 °
  • Bar graph and numeric digital display
  • Meets or exceeds DIN /ISO standards
  • Maximum error on .0005" scale at full range: .0004"
  • .080" contact ball diameter
SPECS & DESCRIPTION
Specifications & Description
  • Description1/2\" Electronic Test Indicator (Depth Gauges/Indicators)
Key Features
  • 4 measuring modes: Min, Max, TIR, and Direct
  • True inch / metric conversion
  • Water, oil and coolant resistant to IP65
  • 1-1/2" diameter dial face rotates a full 360 °
  • Bar graph and numeric digital display
  • Meets or exceeds DIN /ISO standards
  • Maximum error on .0005" scale at full range: .0004"
  • .080" contact ball diameter