Fowler 54-825-200 Clino 2000-Prec. Inclinometer
Representative image only.
Fowler Mfr # 54-825-200 – Item # EW-97604-14
Ideal for the highest precision-levelling and flatness-measuring tasks
  • Highest possible precision over large measuring range of ±45° with integrated temperature compensation
  • Effortless zero adjustment by using the integrated software and a reversal measurement
  • Easy to calibrate, due to implemented software guidance and the calibration aids included
  • State of the art digital technology
  • Built-in possibility to connect an additional instrument for differential measurement or Zerotronic sensors by using the serial I/O port
  • Large digital display for setting all commonly used measuring units
  • The instrument is fully compatible with the whole range of digital sensors from Wyler
  • Powered by 1.5 V batteries, rechargeable batteries or AC adapter
  • Fulfills the strict European CE requirements (immunity against electromagnetic noise)
  • Rugged stainless steel housing with prismatic bases allows use on flat or cylindrical surfaces
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionClino 2000-Prec. Inclinometer
  • Warranty1 year
1 Year Warranty
Key Features
  • Highest possible precision over large measuring range of ±45° with integrated temperature compensation
  • Effortless zero adjustment by using the integrated software and a reversal measurement
  • Easy to calibrate, due to implemented software guidance and the calibration aids included
  • State of the art digital technology
  • Built-in possibility to connect an additional instrument for differential measurement or Zerotronic sensors by using the serial I/O port
  • Large digital display for setting all commonly used measuring units
  • The instrument is fully compatible with the whole range of digital sensors from Wyler
  • Powered by 1.5 V batteries, rechargeable batteries or AC adapter
  • Fulfills the strict European CE requirements (immunity against electromagnetic noise)
  • Rugged stainless steel housing with prismatic bases allows use on flat or cylindrical surfaces
  • 1 year warranty
More About this Item
Our new digital inclination measuring instrument combines precision and quality for a great variety of measuring tasks that will fulfill all your requirements. Ideal for the highest precision leveling and flatness measuring tasks, the unit is a self contained system unparalleled in precision, accuracy and measuring range.