Fungilab V12Z26 Silicone Rotational Viscosity Standard Oil, 500 mPa*s; 600 mL/bottle
Representative image only.
Cole-Parmer Mfr # V12Z26 – Item # EW-98965-19
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Viscosity501.9 mPa*s @25C
  • MaterialSilicone oil
  • DescriptionSilicone Rotational Viscosity Standard Oil, 500 mPa*s; 600 mL/bottle
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Expand testing capabilities and accuracy with these viscometer parts and accessories.
Ensure accurate viscosity readings by calibrating your viscometer to a known viscosity standard.