GE 81001B CO2 Sensor No Display Black Ventostat Wall Mount
Representative image only.
Amphenol Mfr # T8100B – Item # EW-10101-42
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCO2 Sensor No Display Black Ventostat Wall Mount
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
FEATURES
  • Gas sensing engine provides high accuracy in a compact low cost package
  • ABC Logic™ self-calibration system eliminates the need for manual calibration in most applications
  • Gold plated optical sensor increases sensor life and durability
  • Mounting bracket with terminal block provides quick, easy wiring
  • Dual simultaneous analog output (V & mA)
  • On board relay with adjustable set point and dead band
  • Choice of 9 preprogrammed ''standard settings'' on easy to use interface
  • PC interface and adjustable settings allows for simple configuraiton
  • Optional enclosures available
  • Lifetime calibration guarantee