GE Druck 800P2-3000-300-G Dual Pressure Indicator, 3000 & 300 PSIG, 0.01% FS
Representative image only.
GE Druck Mfr # I800PD-2-3000-300-G – Item # EW-68889-53
  • Dual Range: 3000 and 300 psig
  • Accuracy: 0.01% full scale
  • 1 year warranty
$2,775.00USD / Each
sign in for your price.
$216.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDual Pressure Indicator, 3000 & 300 PSIG, 0.01% FS
  • Warranty1 Year
Key Features
  • Dual Range: 3000 and 300 psig
  • Accuracy: 0.01% full scale
  • 1 year warranty
More About this Item
Maximum versatility and range are provided by the DPI 800 series pressure instruments. You can choose a single- or dual-range instrument with a standard or precision range that best fits your needs.
These instruments are also compatible via plug-and-play connection with a range of universal external pressure modules that provides even more flexibility.