GE Druck DPI 705 Portable Pressure Indicator with Internal Sensor, 0 to 30 psia; 1/8" NPT(F)
Representative image only.
GE Druck Mfr # I705-30PSIA – Item # EW-68970-60
  • Audiovisual alarm alerts you once high setpoint is reached--perfect for maintenance and trouble shooting.
  • NEMA 12(IP54) case--ideal for field use.
  • View minimum and maximum readings as well as ambient temperature.
  • Recalibrate using zero and TARE keys.
  • Automatic shut-off and low-battery indication.
  • Perform a one-minute LEAK test with count-down timer displayed.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.1% full-scale
  • Min Pressure (psia)0
  • Max Pressure (psia)30
  • Process Connection1/8\" NPT(F)
  • Unit Of Measurementpsi, \" WC, ft WC, m WC, \" Hg, mm Hg, m Hg, mbar, bar, Pa, hPa, kPa, MPa, kg/cm², and user-definable scale
  • Display5-digit LCD, 0.5\" H
  • Battery TypeThree AA (included)
  • Battery Life300 hours, average
  • Media CompatibilityClean, inert, noncorrosive gases and liquids
  • Resolution0.001 psi
  • DescriptionPortable Pressure Indicator with Internal Sensor, 0 to 30 psia; 1/8\" NPT(F)
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Audiovisual alarm alerts you once high setpoint is reached--perfect for maintenance and trouble shooting.
  • NEMA 12(IP54) case--ideal for field use.
  • View minimum and maximum readings as well as ambient temperature.
  • Recalibrate using zero and TARE keys.
  • Automatic shut-off and low-battery indication.
  • Perform a one-minute LEAK test with count-down timer displayed.