GE Druck IDT 600-2 Dirt/Moisture Trap for Calibrators
Representative image only.
GE Druck Mfr # IDT600-2 – Item # EW-68452-26
Prevent Contamination of your Pneumatic Calibrator
  • Easy to install and clean
  • Does not affect calibration accuracy
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDirt/Moisture Trap for Calibrators
  • Warranty1 Year
1 Year Warranty
Key Features
  • Easy to install and clean
  • Does not affect calibration accuracy
  • 1 year warranty
More About this Item
The unit is an effective barrier against the transfer of moisture and dirt from an instrument under test to any portable pressure calibrator. Installed directly on the calibrator pressure port, regular use of this trap will prevent contamination of the calibrator's pneumatic system, improving long-term reliability and reducing maintenance costs. The trap has no diaphragm or other internal components which could create a pressure drop and does not affect calibration accuracy. The unit is simple to dismantle and easy to clean. It should be periodically cleaned to prevent excessive contaminate build-up which may overflow into the calibrator. The top and bottom sections are simply unscrewed, releasing the transparent chamber which is then wiped clean.