GE Druck IO620-CASE-2 Protective System Carrying Case
Representative image only.
GE Druck Mfr # IO620-CASE-2 – Item # EW-23001-52
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionProtective System Carrying Case
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Intecal Field Calibration Manager
This software module adds the general features of Intecal calibration management software to the DPI 620 allowing new device records and procedures to be created in the field while giving access to full historical data. It works seamlessly with the documenting capability of the DPI 620 to automate field calibration and fully integrates with the master database of Intecal.
Carrying case
A protective fabric carrying case with belt loop, shoulder strap and large pocket for test leads and accessories.
System carrying case
Attaché style carrying case for system components including the DPI 620, MC 620, PM 620 modules, test leads, hose and adaptors.
Pressure station carrying case
A