GE Druck IPM620S-008G IS Pressure Module for IS DPI 620 (+/-10 inH2O)
Representative image only.
GE Druck Mfr # IPM620S-008G – Item # EW-50015-07
Acts as stand-alone pressure generator
  • Replaces conventional hand pumps
  • Can be used on workbench as comparators
  • Offers operating temperature of 14 to 104°F (-10 tp 40°C)
PRODUCT OPTIONS
$2,523.00USD / Each
sign in for your price.
$199.00USD / Each
for an additional
$199.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionIS Pressure Module for IS DPI 620 ( /-10 inH2O)
Key Features
  • Replaces conventional hand pumps
  • Can be used on workbench as comparators
  • Offers operating temperature of 14 to 104°F (-10 tp 40°C)
More About this Item
Pressure Stations
If pressure generation is required, the calibrator and pressure module can be attached to one of three pressure generation stations, to form a fully-integrated pressure calibrator of unparalleled performance. Each pressure station is designed for stand-alone operation as a pressure generator and can replace conventional hand pumps to provide greater efficiency, ease of use and significantly reduce the risk of repetitive strain injuries. They can also be used on the workbench as comparators.
Pressure Modules
The pressure modules can be used in conjunction with the relevant pneumatic or hydraulic pressure station and the DPI 620 multifunction calibrator to form an integrated pressure calibrator for test and calibration of pressure instruments including transmitters, transducers, switches, gauges, indicators and recorders. Alternatively, the modules can be combined with the pressure station by a module carrier (MC620), which adds a dual channel pressure measurement capability that can be re-ranged in seconds.
The modules are fully interchangeable—no need for set-up or calibration. They offer simple screw fit—no tools required. Ranges from 10 in H2O to 15,000 psi with a total measurement uncertainty, including operation over -10 to 40°C (14 to 104°F) for one year. Accuracy from 0.005% FS.